RVX-208 99%

Supplier: Ambeed
Warning

Synonyms: Apabetalone, 2-(4-(2-Hydroxyethoxy)-3,5-dimethylphenyl)-5,7-dimethoxyquinazolin-4(3H)-one

A161440-10MG A161440-1MG A161440-50MG A161440-5MG A161440-1G
76872-020EA 13.8 USD
76872-020 76872-022 76872-024 76872-026 77671-282
RVX-208 99%
RVX-208

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₅
MW: 370.4 g/mol
MDL Number: MFCD18633270
CAS Number: 1044870-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR