Vernakalant hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: Vernakalant HCl, RSD1235 HCl, (R)-1-((1R,2R)-2-(3,4-Dimethoxyphenethoxy)cyclohexyl)pyrrolidin-3-ol hydrochloride

A161054-10MG A161054-1MG A161054-25MG A161054-2MG A161054-50MG A161054-5MG
76872-004EA 43 USD
76872-004 76872-006 76872-008 76872-010 76872-012 76872-014
Vernakalant hydrochloride 98+%
Vernakalant hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₃₂ClNO₄
MDL Number: MFCD09833303
CAS Number: 748810-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR