Methyl 4-Boc-morpholine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butyl 2-methyl morpholine-2,4-dicarboxylate

A160577-100MG A160577-1G A160577-250MG A160577-5G
76871-992EA 15 USD
76871-992 76871-994 76871-996 76871-998
Methyl 4-Boc-morpholine-2-carboxylate 97%
Methyl 4-Boc-morpholine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD16877230
CAS Number: 500789-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR