Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate 97%
Supplier: Ambeed
|
Warning
|
A160549-100MG
A160549-1G
A160549-250MG
A160549-5G
76871-984EA
21.22
USD
76871-984
76871-986
76871-988
76871-990
Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate 97%
Diphenyl(trifluoromethyl)sulfonium trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₁₄H₁₀F₆O₃S₂ MW: 404.35 g/mol |
MDL Number:
MFCD12912173 CAS Number: 147531-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...