6-Chloro-4-methylpyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A159171-100MG A159171-10G A159171-1G A159171-250MG A159171-5G
76871-962EA 13.8 USD
76871-962 76871-964 76871-966 76871-968 77592-938
6-Chloro-4-methylpyridazin-3(2H)-one 97%
6-Chloro-4-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD13659416
CAS Number: 1834-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR