Di-tert-butyl fumarate 98%

Supplier: Ambeed
Warning

A158233-1G A158233-250MG
76871-946EA 87 USD
76871-946 76871-948
Di-tert-butyl fumarate 98%
Di-tert-butyl fumarate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MW: 228.28 g/mol
MDL Number: MFCD23701540
CAS Number: 7633-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR