(7-Methoxy-1-naphthyl)acetic acid 98%

Supplier: Ambeed
Warning

A156414-1G A156414-25G
76871-878EA 9 USD
76871-878 76871-880
(7-Methoxy-1-naphthyl)acetic acid 98%
(7-Methoxy-1-naphthyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
MDL Number: MFCD12756806
CAS Number: 6836-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR