1,4,7-Tritosyl-1,4,7-triazonane 97%

Supplier: Ambeed
Warning

A155984-100MG A155984-250MG A155984-1G
76871-860EA 15.92 USD
76871-860 76871-862 77592-916
1,4,7-Tritosyl-1,4,7-triazonane 97%
1,4,7-Tritosyl-1,4,7-triazonane

Precaución: for Research Use Only

Formula: C₂₇H₃₃N₃O₆S₃
MW: 591.76 g/mol
MDL Number: MFCD00184056
CAS Number: 52667-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR