Ethyl 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A155865-10G A155865-1G A155865-250MG A155865-25G A155865-5G
76871-846EA 94.43 USD
76871-846 76871-848 76871-850 76871-852 76871-854
Ethyl 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate 97%
Ethyl 1-(4-methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₂₇H₂₈N₄O₅
MW: 488.53 g/mol
MDL Number: MFCD18072444
CAS Number: 503614-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR