5-Chloro-2-iodoanisole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-iodophenyl methyl ether, 4-Chloro-1-iodo-2-methoxybenzene

A155312-100MG A155312-1G A155312-250MG A155312-5G
76871-816EA 6 USD
76871-816 76871-818 76871-820 76871-822
5-Chloro-2-iodoanisole 95%
5-Chloro-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆ClIO
MDL Number: MFCD09753735
CAS Number: 755027-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR