(S)-Piperidin-3-ylmethanol hydrochloride 95%

Supplier: Ambeed
Warning

A155160-1G A155160-25G
76871-812EA 29.71 USD
76871-812 76871-814
(S)-Piperidin-3-ylmethanol hydrochloride 95%
(S)-Piperidin-3-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
MDL Number: MFCD14155681
CAS Number: 1125551-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR