2-Methyl-4-(2-methylbenzamido)benzoic acid 97%

Supplier: Ambeed
Warning

A155131-100G A155131-10G A155131-1G A155131-25G A155131-5G
76871-802EA 501 USD
76871-802 76871-804 76871-806 76871-808 76871-810
2-Methyl-4-(2-methylbenzamido)benzoic acid 97%
2-Methyl-4-(2-methylbenzamido)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃
MW: 269.29 g/mol
MDL Number: MFCD16038736
CAS Number: 317374-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR