4-Iodo-2-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A154560-10G A154560-1G A154560-250MG A154560-5G
76871-786EA 168.69 USD
76871-786 76871-788 76871-790 77611-466
4-Iodo-2-(trifluoromethyl)benzoic acid 95%
4-Iodo-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃IO₂
MW: 316.016 g/mol
MDL Number: MFCD09881756
CAS Number: 954815-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR