N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine 98%

Supplier: Ambeed
Warning

A154254-1G A154254-25G A154254-5G
76871-784EA 12 USD
76871-784 77883-210 77883-211
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine 98%
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₄
MDL Number: MFCD00076900
CAS Number: 76985-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR