Amino- PEG3-t-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: NH2-PEG3-C2-Boc, tert-Butyl 3-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)propanoate

A154148-1G A154148-250MG A154148-25G A154148-5G A154148-100MG
76871-772EA 23 USD
76871-772 76871-774 76871-776 77567-244 77671-168
Amino- PEG3-t-butyl ester 97%
Amino- PEG3-t-butyl ester

Precaución: For research use only.

Formula: C₁₃H₂₇NO₅
MDL Number: MFCD06201017
CAS Number: 252881-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR