5-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-Dioxaborolane, 2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-, 5-Fluoro-2-methoxyphenylboronic acid pinacol ester

A154013-1G A154013-250MG A154013-5G
76871-766EA 21 USD
76871-766 76871-768 76871-770
5-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%
5-Fluoro-2-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MDL Number: MFCD16996296
CAS Number: 1383806-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR