tert-Butyl (2-(benzylamino)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A153751-1G A153751-250MG A153751-5G A153751-10G A153751-100MG
76871-746EA 18 USD
76871-746 76871-748 76871-750 77611-454 77671-158
tert-Butyl (2-(benzylamino)ethyl)carbamate 95%
tert-Butyl (2-(benzylamino)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₂
MDL Number: MFCD04114275
CAS Number: 174799-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR