(S)-tert-butyl (piperidin-3-ylmethyl)carbamate 97%

Supplier: Ambeed
Warning

A152950-1G A152950-250MG A152950-5G
76871-696EA 24 USD
76871-696 76871-698 76871-700
(S)-tert-butyl (piperidin-3-ylmethyl)carbamate 97%
(S)-tert-butyl (piperidin-3-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD03093385
CAS Number: 1016167-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR