2-Bromo-4-fluoro-5-methylbenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A152893-100G
A152893-10G
A152893-250MG
A152893-25G
A152893-5G
76871-686EA
387
USD
76871-686
76871-688
76871-690
76871-692
76871-694
2-Bromo-4-fluoro-5-methylbenzonitrile 98%
2-Bromo-4-fluoro-5-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅BrFN |
MDL Number:
MFCD07782070 CAS Number: 916792-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...