2'-Anilino-6'-(dibutylamino)-3'-methylfluoran 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6'-(Dibutylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 3'-Dibutylamino-6'-methyl-7'-phenylaminofluoran, ODB 2
A152356-100G
A152356-1G
A152356-25G
A152356-5G
A152356-500G
76871-650EA
132
USD
76871-650
76871-652
76871-654
76871-656
77883-192
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran 97%
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran
Precaución: for Research Use Only
Formula:
C₃₅H₃₆N₂O₃ |
MDL Number:
MFCD00307579 CAS Number: 89331-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...