1-Hydroxy-6-oxo-1,6-dihydropyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A151742-100MG A151742-1G A151742-250MG
76871-626EA 50.93 USD
76871-626 76871-628 77611-416
1-Hydroxy-6-oxo-1,6-dihydropyridine-2-carboxylic acid 97%
1-Hydroxy-6-oxo-1,6-dihydropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₄
MW: 155.11 g/mol
MDL Number: MFCD20270107
CAS Number: 94781-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR