((R)-Piperidin-2-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: (R)-Piperidin-2-ylmethanol

A150533-100MG A150533-1G A150533-250MG A150533-25G
76871-602EA 18.04 USD
76871-602 76871-604 76871-606 76871-608
((R)-Piperidin-2-yl)methanol 97%
((R)-Piperidin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD11036292
CAS Number: 3197-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR