2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-tert-butyl ester 97%
Supplier: Ambeed
|
Warning
|
A150446-100MG
A150446-1G
A150446-250MG
A150446-5G
A150446-25G
76871-588EA
34
USD
76871-588
76871-590
76871-592
76871-594
77671-118
2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-tert-butyl ester 97%
2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₉NO₄ |
MDL Number:
MFCD17016194 CAS Number: 1211526-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...