7-Iodoquinazoline-2,4(1H,3H)-dione 96%

Supplier: Ambeed
Warning

A150193-1G A150193-250MG
76871-584EA 207.94 USD
76871-584 76871-586
7-Iodoquinazoline-2,4(1H,3H)-dione 96%
7-Iodoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅IN₂O₂
MW: 288.04 g/mol
MDL Number: MFCD09954830
CAS Number: 959236-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR