4-Chloronicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-cyanopyridine

A149566-100MG A149566-1G A149566-250MG A149566-5G
76871-558EA 13.8 USD
76871-558 76871-560 76871-562 76871-564
4-Chloronicotinonitrile 95%
4-Chloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
MW: 138.5544 g/mol
MDL Number: MFCD08234759
CAS Number: 89284-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR