2-Ethylbenzyl alcohol 97%

Supplier: Ambeed
Warning

A149445-10G A149445-1G A149445-250MG A149445-5G A149445-25G
76871-550EA 129 USD
76871-550 76871-552 76871-554 76871-556 77671-106
2-Ethylbenzyl alcohol 97%
2-Ethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD03093889
CAS Number: 767-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR