2,3-Difluoro-6-nitrophenol 98%

Supplier: Ambeed
Danger

A149195-100G A149195-10G A149195-25G A149195-500G A149195-5G
76871-534EA 74 USD
76871-534 76871-536 76871-538 76871-540 76871-542
2,3-Difluoro-6-nitrophenol 98%
2,3-Difluoro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MDL Number: MFCD00042255
CAS Number: 82419-26-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR