4-Bromo-1H-imidazole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A149184-1G A149184-250MG
76871-530EA 371 USD
76871-530 76871-532
4-Bromo-1H-imidazole-2-carboxylic acid 95%
4-Bromo-1H-imidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
MDL Number: MFCD09842486
CAS Number: 1260753-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR