2-Amino-4-fluoro-5-methylbenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A148158-1G
A148158-250MG
A148158-5G
76871-502EA
42
USD
76871-502
76871-504
76871-506
2-Amino-4-fluoro-5-methylbenzonitrile 97%
2-Amino-4-fluoro-5-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₇FN₂ |
MDL Number:
MFCD15529743 CAS Number: 1357942-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...