8-Ethoxy-5-nitroquinoline 98%

Supplier: Ambeed
Warning

A147013-1G A147013-250MG
76871-474EA 181.42 USD
76871-474 76871-476
8-Ethoxy-5-nitroquinoline 98%
8-Ethoxy-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₃
MW: 218.2087 g/mol
MDL Number: MFCD00777819
CAS Number: 19746-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR