N3-Benzylpyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A146605-1G A146605-250MG
76871-466EA 83.82 USD
76871-466 76871-468
N3-Benzylpyridine-3,4-diamine 95%
N3-Benzylpyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃
MW: 199.25 g/mol
MDL Number: MFCD11114581
CAS Number: 75115-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR