4,6-Diaminopyrimidine-2-thiol 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Diamino-2-mercaptopyrimidine, 4,6-Diamino-2-pyrimidinethiol

A145858-100G A145858-10G A145858-1G A145858-25G A145858-500G A145858-5G
76871-448EA 177.18 USD
76871-448 76871-450 76871-452 76871-454 76871-456 76871-458
4,6-Diaminopyrimidine-2-thiol 97%
4,6-Diaminopyrimidine-2-thiol

Precaución: for Research Use Only

Formula: C₄H₆N₄S
MW: 142.18 g/mol
MDL Number: MFCD00006078
CAS Number: 1004-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR