2,5-Dihydroxypyridine 98%

Supplier: Ambeed
Warning

A145230-100MG A145230-1G A145230-250MG A145230-25G A145230-5G
76871-420EA 8 USD
76871-420 76871-422 76871-424 76871-426 77883-161
2,5-Dihydroxypyridine 98%
2,5-Dihydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MDL Number: MFCD00234075
CAS Number: 5154-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR