1-Phenylpiperidin-2-one 98%

Supplier: Ambeed
Warning

A145029-1G A145029-250MG A145029-100MG
76871-416EA 166 USD
76871-416 76871-418 77883-160
1-Phenylpiperidin-2-one 98%

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MDL Number: MFCD04037163
CAS Number: 4789-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR