2-Amino-5-methyl-3-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-5-nitro-3-picoline, 2-Amino-3-nitro-5-picoline, 5-Methyl-3-nitro-2-pyridylamine, 5-Methyl-3-nitropyridin-2-amine

A144613-10G A144613-1G A144613-250MG A144613-25G A144613-5G
76871-402EA 17 USD
76871-402 76871-404 76871-406 76871-408 76871-410
2-Amino-5-methyl-3-nitropyridine 97%
2-Amino-5-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14
MDL Number: MFCD00047442
CAS Number: 7598-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR