3-O-ethyl-L-ascorbic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-5-((S)-1,2-Dihydroxyethyl)-4-ethoxy-3-hydroxyfuran-2(5H)-one
A144601-10G
A144601-1G
A144601-25G
A144601-5G
76871-394EA
18.04
USD
76871-394
76871-396
76871-398
76871-400
3-O-ethyl-L-ascorbic acid 95%
3-O-ethyl-L-ascorbic acid
Precaución: for Research Use Only
Formula:
C₈H₁₂O₆ MW: 204.18 g/mol |
MDL Number:
MFCD09261382 CAS Number: 86404-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...