4,6-Dimethoxy-5-nitropyrimidine 97%

Supplier: Ambeed
Warning

A144538-10G A144538-1G A144538-5G
76871-386EA 156 USD
76871-386 76871-388 76871-390
4,6-Dimethoxy-5-nitropyrimidine 97%
4,6-Dimethoxy-5-nitropyrimidine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄
MDL Number: MFCD00129731
CAS Number: 15846-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR