Ethyl-4-chloro-6,8-difluoroquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

A144420-100MG A144420-250MG A144420-5G
76871-380EA 28.65 USD
76871-380 76871-382 76871-384
Ethyl-4-chloro-6,8-difluoroquinoline-3-carboxylate 95%
Ethyl 4-chloro-6,8-difluoroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₈ClF₂NO₂
MW: 271.65 g/mol
MDL Number: MFCD00173353
CAS Number: 150258-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR