3-Azaspiro[5.5]undecane hydrochloride 98%

Supplier: Ambeed
Warning

A144342-100MG A144342-250MG A144342-5G A144342-1G
76871-374EA 29 USD
76871-374 76871-376 76871-378 77611-328
3-Azaspiro[5.5]undecane hydrochloride 98%
3-Azaspiro[5.5]undecane hydrochloride

Precaución: for Research Use Only

Formula: C10H20ClN
MDL Number: MFCD02093801
CAS Number: 1125-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR