8-Iodoquinazolin-4(3H)-one 98%

Supplier: Ambeed
Warning

A144217-100MG A144217-1G A144217-250MG A144217-5G
76871-360EA 18 USD
76871-360 76871-362 76871-364 76871-366
8-Iodoquinazolin-4(3H)-one 98%
8-Iodoquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅IN₂O
MDL Number: MFCD09954852
CAS Number: 77150-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR