4,6-Diaminopyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Pyrimidinediamine

A143419-100G A143419-10G A143419-1G A143419-25G A143419-5G
76871-324EA 337.37 USD
76871-324 76871-326 76871-328 76871-330 76871-332
4,6-Diaminopyrimidine 97%
4,6-Diaminopyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD00044572
CAS Number: 2434-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR