Methyl 2-fluoro-3-methylbenzoate 97%

Supplier: Ambeed
Warning

A143405-100G A143405-100MG A143405-10G A143405-1G A143405-250MG A143405-25G A143405-5G
76871-310EA 531 USD
76871-310 76871-312 76871-314 76871-316 76871-318 76871-320 76871-322
Methyl 2-fluoro-3-methylbenzoate 97%
Methyl 2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD10698751
CAS Number: 586374-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR