5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-carboxamide

A142739-100MG A142739-1G A142739-250MG
76871-294EA 8 USD
76871-294 76871-296 76871-298
5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide 97%
5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₉H₁₄N₄O₂
MW: 210.233 g/mol
MDL Number: MFCD12405839
CAS Number: 1082745-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR