N-Boc-L-alaninol 98%

Supplier: Ambeed
Warning

Synonyms: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

A142574-1000G A142574-100G A142574-10G A142574-1G A142574-25G A142574-500G A142574-5G
76871-280EA 453.01 USD
76871-280 76871-282 76871-284 76871-286 76871-288 76871-290 76871-292
N-Boc-L-alaninol 98%
N-Boc-L-alaninol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MW: 175.2 g/mol
MDL Number: MFCD00043121
CAS Number: 79069-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR