Methyl-2-(3-aminophenyl)acetate hydrochloride 98%

Supplier: Ambeed
Warning

A142008-1G A142008-5G
76871-260EA 28 USD
76871-260 76871-262
Methyl-2-(3-aminophenyl)acetate hydrochloride 98%
Methyl-2-(3-aminophenyl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MDL Number: MFCD11501071
CAS Number: 150319-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR