N-(Benzyloxy)-2-nitrobenzenesulfonamide 95%

Supplier: Ambeed
Danger

A141569-10G A141569-1G A141569-250MG A141569-5G
76871-238EA 149 USD
76871-238 76871-240 76871-242 76871-244
N-(Benzyloxy)-2-nitrobenzenesulfonamide 95%
N-(Benzyloxy)-2-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₅S
MDL Number: MFCD23135238
CAS Number: 77925-80-5
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR