(2-Amino-5-fluorophenyl)methanol 95%

Supplier: Ambeed
Warning

A141520-1G A141520-250MG A141520-100MG A141520-5G
76871-234EA 33 USD
76871-234 76871-236 77883-132 77883-133
(2-Amino-5-fluorophenyl)methanol 95%
(2-Amino-5-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MDL Number: MFCD08234510
CAS Number: 748805-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR