1-(3-Bromoquinolin-6-yl)ethanone 97%

Supplier: Ambeed
Warning

A141420-100MG A141420-250MG
76871-218EA 5 USD
76871-218 76871-220
1-(3-Bromoquinolin-6-yl)ethanone 97%
1-(3-Bromoquinolin-6-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MDL Number: MFCD12024483
CAS Number: 1150618-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR