2,6-Dibromo-3-fluoropyridine 95%
Supplier: Ambeed
|
Warning
|
A140897-100MG
A140897-250MG
A140897-1G
A140897-10G
76871-206EA
21
USD
76871-206
76871-208
77611-284
77883-125
2,6-Dibromo-3-fluoropyridine 95%
2,6-Dibromo-3-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₂Br₂FN |
MDL Number:
MFCD13185513 CAS Number: 41404-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...