Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Ser-OMe

A140799-100G A140799-10G A140799-1G A140799-25G A140799-5G
76871-192EA 125.19 USD
76871-192 76871-194 76871-196 76871-198 76871-200
Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 97%
Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.23 g/mol
MDL Number: MFCD03756651
CAS Number: 69942-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR